missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S6K2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15268-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains a kinase catalytic domain and phosphorylates the S6 ribosomal protein and eukaryotic translation initiation factor 4B (eIF4B). Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation.Specifications
| S6K2 | |
| Polyclonal | |
| Unconjugated | |
| RPS6KB2 | |
| KLS, P70 beta, P70 beta 1, P70 beta 2, p70 ribosomal S6 kinase beta, P70 S6 kinase beta, p70 S6K 2, p70 S6K beta, p70 S6KB, p70(S6K) beta, P70S6K2, p70S6Kb, RPS6KB2, S6 kinase related kinase, S6K beta, S6K beta 2, S6K beta2, S6K2, SRK, STK14B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361696, 58988, 6199 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UBS0, Q9Z1M4 | |
| Rps6kb2 | |
| S6K2 Fusion Protein Ag7371 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction