Learn More
(S)-Terfenadine N-Oxide, Mikromol™
Description
(S)-Terfenadine N-Oxide
Specifications
Specifications
| Chemical Name or Material | (S)-Terfenadine N-Oxide |
| CAS | 2732309-85-0 |
| Analyte or Component Names | (S)-Terfenadine N-Oxide |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C32 H41 N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-Piperidinebutanol, α-[4-(1,1-dimethylethyl)phenyl]-4-(hydroxydiphenylmethyl)-, 1-oxide |
| InChI Formula | InChI=1S/C32H41NO3/c1-31(2,3)26-18-16-25(17-19-26)30(34)15-10-22-33(36)23-20-29(21-24-33)32(35,27-11-6-4-7-12-27)28-13-8-5-9-14-28/h4-9,11-14,16-19,29-30,34-35H,10,15,20-24H2,1-3H3/t29?,30-,33?/m0/s1 |
| SMILES | CC(C)(C)c1ccc(cc1)[C@@H](O)CCC[N+]2([O-])CCC(CC2)C(O)(c3ccccc3)c4ccccc4 |
| IUPAC Name | (1S)-1-(4-tert-butylphenyl)-4-[4-[hydroxy(diphenyl)methyl]-1-oxidopiperidin-1-ium-1-yl]butan-1-ol |
| Show More |