Learn More
(S)-(-)-Carvedilol, TRC
Product Code. 30134922
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134922 50 mg 50mg
17984845 10 mg 10mg
17924305 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134922 Supplier Toronto Research Chemicals Supplier No. TRCC18463550MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 95094-00-1
Molecular Formula C24H26N2O4
Molecular Weight (g/mol) 406.47
Synonym 2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-, (2S)- (9CI), 2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-, (S)-, (-)-Carvedilol, (S)-(-)-Carvedilol, (S)-Carvedilol
IUPAC Name (2S)-1-(9H-carbazol-4-yloxy)-3-[2-(2-methoxyphenoxy)ethylamino]propan-2-ol
SMILES COc1ccccc1OCCNC[C@H](O)COc2cccc3[nH]c4ccccc4c23
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C24H26N2O4/c1-28-21-10-4-5-11-22(21)29-14-13-25-15-17(27)16-30-23-12-6-9-20-24(23)18-7-2-3-8-19(18)26-20/h2-12,17,25-27H,13-16H2,1H3/t17-/m0/s1
Formula Weight 406.1893
Chemical Name or Material (S)-(-)-Carvedilol