CAS RN 95094-00-1
CAS RN 95094-00-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-(-)-Carvedilol, TRC
CAS: 95094-00-1 Molecular Formula: C24H26N2O4 Molecular Weight (g/mol): 406.47 Synonym: 2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-, (2S)- (9CI),2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-, (S)-,(-)-Carvedilol,(S)-(-)-Carvedilol,(S)-Carvedilol IUPAC Name: (2S)-1-(9H-carbazol-4-yloxy)-3-[2-(2-methoxyphenoxy)ethylamino]propan-2-ol SMILES: COc1ccccc1OCCNC[C@H](O)COc2cccc3[nH]c4ccccc4c23