Learn More
(S)-(+)-3-Chloro-1,2-propanediol, 98%, ee 98%
Product Code. 11369405
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11369405
Brand: Thermo Scientific Alfa Aesar L18556.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 60827-45-4 | C3H7ClO2 | 110.537 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 60827-45-4
Molecular Formula C3H7ClO2
Molecular Weight (g/mol) 110.537
MDL Number MFCD00210270
InChI Key SSZWWUDQMAHNAQ-GSVOUGTGSA-N
Synonym s-+-3-chloro-1,2-propanediol, s-3-chloro-1,2-propanediol, s-3-chloropropane-1,2-diol, 2s-3-chloropropane-1,2-diol, s-alpha-chlorohydrin, unii-227bd3h6h6, ccris 7386, 1,2-propanediol, 3-chloro-, s, 1,2-propanediol, 3-chloro-, 2s
PubChem CID 148904
IUPAC Name (2S)-3-chloropropane-1,2-diol
SMILES C(C(CCl)O)O
Specifications
Density 1.322
Boiling Point 213°C
Flash Point 126°C (258°F)
Refractive Index 1.48
Quantity 25 g
UN Number UN2689
Beilstein 2202929
Optical Rotation +0.9° (Neat)
Formula Weight 110.54
Percent Purity 98%
Assay ee 98%
Chemical Name or Material (S)-(+)-3-Chloro-1,2-propanediol