CAS RN 60827-45-4
CAS RN 60827-45-4
IUPAC Name:
(2S)-3-chloropropane-1,2-diol
Synonyms:
(2S)-3-chloropropane-1,2-diol
Molecular Weight (g/mol):
110.54
Molecular Formula:
C3H7ClO2
InChi Key:
SSZWWUDQMAHNAQ-GSVOUGTGSA-N
SMILES:
OC[C@H](O)CCl
1
–
1
of
1
results
1
(S)-(+)-3-Chloro-1,2-propanediol, 98%, ee 98%
CAS: 60827-45-4 Molecular Formula: C3H7ClO2 Molecular Weight (g/mol): 110.537 MDL Number: MFCD00210270 InChI Key: SSZWWUDQMAHNAQ-GSVOUGTGSA-N Synonym: s-+-3-chloro-1,2-propanediol,s-3-chloro-1,2-propanediol,s-3-chloropropane-1,2-diol,2s-3-chloropropane-1,2-diol,s-alpha-chlorohydrin,unii-227bd3h6h6,ccris 7386,1,2-propanediol, 3-chloro-, s,1,2-propanediol, 3-chloro-, 2s PubChem CID: 148904 IUPAC Name: (2S)-3-chloropropane-1,2-diol SMILES: C(C(CCl)O)O