missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Chloro-1,2-propanediol, 98%, ee 98%

Product Code. 11349405
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11349405 1 g 1g
11359405 5 g 5g
11369405 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11349405

Brand: Thermo Scientific Alfa Aesar L18556.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 60827-45-4 | C3H7ClO2 | 110.537 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60827-45-4
Molecular Formula C3H7ClO2
Molecular Weight (g/mol) 110.537
MDL Number MFCD00210270
InChI Key SSZWWUDQMAHNAQ-GSVOUGTGSA-N
Synonym s-+-3-chloro-1,2-propanediol, s-3-chloro-1,2-propanediol, s-3-chloropropane-1,2-diol, 2s-3-chloropropane-1,2-diol, s-alpha-chlorohydrin, unii-227bd3h6h6, ccris 7386, 1,2-propanediol, 3-chloro-, s, 1,2-propanediol, 3-chloro-, 2s
PubChem CID 148904
IUPAC Name (2S)-3-chloropropane-1,2-diol
SMILES C(C(CCl)O)O

Specifications

Density 1.322
Boiling Point 213°C
Flash Point 126°C (258°F)
Refractive Index 1.48
Quantity 1 g
UN Number UN2689
Beilstein 2202929
Optical Rotation +0.9° (Neat)
Formula Weight 110.54
Percent Purity 98%
Assay ee 98%
Chemical Name or Material (S)-(+)-3-Chloro-1,2-propanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.