Learn More
(S)-(-)-3-Amino-1,2-propanediol, 98%
Product Code. p-7029296
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11302739 1 g 1g
11312739 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11302739 Supplier Thermo Scientific Alfa Aesar Supplier No. H26743.03
Add to basket
missing translation for 'validMainframeMsg'
CAS: 61278-21-5 | C3H9NO2 | 91.11 g/mol
(S)-(-)-3-Amino-1,2-propanediol is used as a chiral selector ligand, used subsequently for a nucleophilic substitution with the dimethoxytrityl (DMT)-protected
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(-)-3-Amino-1,2-propanediol is used as a chiral selector ligand, used subsequently for a nucleophilic substitution with the dimethoxytrityl (DMT)-protected
Solubility
Soluble in water.
Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 61278-21-5
Molecular Formula C3H9NO2
Molecular Weight (g/mol) 91.11
MDL Number MFCD00798260
InChI Key KQIGMPWTAHJUMN-VKHMYHEASA-N
Synonym s-3-amino-1,2-propanediol, s-3-aminopropane-1,2-diol, s---3-amino-1,2-propanediol, 2s-3-aminopropane-1,2-diol, s---3-amino-1,2-propandiol, 1,2-propanediol, 3-amino-, 2s, ksc496i7p, s-2,3-dihydroxypropylamine, 2s-2,3-dihydroxypropylamine, 2s-3-amino-1,2-propanediol
PubChem CID 440902
ChEBI CHEBI:1456
IUPAC Name (2S)-3-aminopropane-1,2-diol
SMILES C(C(CO)O)N
Specifications
Melting Point 54°C to 56°C
Density 1.175
Boiling Point 117°C to 119°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.483
Quantity 1 g
UN Number UN3259
Beilstein 1719122
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Optical Rotation −28° (c=4, 5M HCl)
Formula Weight 91.11
Percent Purity 98%
Chemical Name or Material (S)-(-)-3-Amino-1,2-propanediol