Learn More
S-[2-(1-Methylethoxy)-2-oxoethyl]-L-cysteine Hydrochloride, Mikromol™
Description
S-[2-(1-Methylethoxy)-2-oxoethyl]-L-cysteine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | S-[2-(1-Methylethoxy)-2-oxoethyl]-L-cysteine Hydrochloride |
| CAS | 2734714-19-1 |
| Analyte or Component Names | S-[2-(1-Methylethoxy)-2-oxoethyl]-L-cysteine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H15 N O4 S . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | (Z)-[5-Fluoro-2-methyl-1-[4-(methylsulfinyl)benzylidene]-1H-inden-3-yl]acetamide, S-[2-(1-Methylethoxy)-2-oxoethyl]-L-cysteine Hydrochloride |
| InChI Formula | InChI=1S/C9H11FN2O5/c1-3-5(13)6(14)8(17-3)12-2-4(10)7(15)11-9(12)16/h2-3,5-6,8,13-14H,1H3,(H,11,15,16)/t3-,5-,6-,8-/m1/s6 |
| SMILES | Cl.CC(C)OC(=O)CSC[C@H](N)C(=O)O |
| IUPAC Name | (2R)-2-amino-3-(2-oxo-2-propan-2-yloxyethyl)sulfanylpropanoic acid;hydrochloride |
| Show More |