missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RS 127445 hydrochloride
GREENER_CHOICE

Product Code. 18044842
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18044842 50 mg 50mg
30001336 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18044842 Supplier Tocris Bioscience™ Supplier No. 2993/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity and selective 5-HT2B antagonist

High affinity 5-HT2B receptor antagonist (pKi = 9.5). Displays 1000-fold selectivity for 5-HT2B with good bioavailability.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199864-86-3
Molecular Formula C17H17ClFN3

Specifications

Quantity 50 mg
Formula Weight 317.79
Purity >98%
Molecular Weight (g/mol) 317.79
Chemical Name or Material RS 127445 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.