accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 199864-86-3

CH3CH3NNFH2NHCl

CAS RN 199864-86-3

IUPAC Name: 4-(4-fluoronaphthalen-1-yl)-6-(propan-2-yl)pyrimidin-2-amine hydrochloride
Synonyms: 4-(4-fluoronaphthalen-1-yl)-6-isopropylpyrimidin-2-amine hydrochloride
Molecular Weight (g/mol): 317.79
Molecular Formula: C17H17ClFN3
InChi Key: MKJPYBJBPRFMHL-UHFFFAOYSA-N
SMILES: Cl.CC(C)C1=NC(N)=NC(=C1)C1=C2C=CC=CC2=C(F)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results