Learn More
Ropinirole N-Oxide Hydrochloride, Mikromol™
Description
Ropinirole N-Oxide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Ropinirole N-Oxide Hydrochloride |
| CAS | 2726492-53-9 |
| Analyte or Component Names | Ropinirole N-Oxide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H24 N2 O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ropinirole N-Oxide Hydrochloride |
| InChI Formula | InChI=1S/C16H24N2O2.ClH/c1-3-9-18(20,10-4-2)11-8-13-6-5-7-15-14(13)12-16(19)17-15;/h5-7H,3-4,8-12H2,1-2H3,(H,17,19);1H |
| SMILES | Cl.CCC[N+]([O-])(CCC)CCc1cccc2NC(=O)Cc12 |
| IUPAC Name | N-[2-(2-oxo-1,3-dihydroindol-4-yl)ethyl]-N-propylpropan-1-amine oxide;hydrochloride |
| Show More |