Learn More
Rolicyprine, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Rolicyprine |
| CAS | 2829-19-8 |
| Analyte or Component Names | Rolicyprine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H16 N2 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-Pyrrolidinecarboxamide, 5-oxo-N-[(1S,2R)-2-phenylcyclopropyl]-, (2S)-, 2-Pyrrolidinecarboxamide, 5-oxo-N-(2-phenylcyclopropyl)-, stereoisomer (8CI), (+)-N-(trans-2-Phenylcyclopropyl)-L-5-pyrrolidone-2-carboxamide, 5-Oxo-N-(D-trans-2-phenylcyclopropyl)-L-2-pyrrolidinecarboxamide, Cypromin, Ex 4883, L-trans-(+)-5-Oxo-N-(2-phenylcyclopropyl)-2-pyrrolidinecarboxamide, RMI 83027, Rolicypram, Rolicyprine, d-5-Oxo-N-(trans-2-phenylcyclopropyl)-2-pyrrolidinecarboxamide |
| InChI Formula | InChI=1S/C14H16N2O2/c17-13-7-6-11(15-13)14(18)16-12-8-10(12)9-4-2-1-3-5-9/h1-5,10-12H,6-8H2,(H,15,17)(H,16,18)/t10-,11+,12+/m1/s1 |
| Solution Type | Neat |
| SMILES | O=C(N[C@H]1C[C@@H]1c2ccccc2)[C@@H]3CCC(=O)N3 |
| Show More |