missing translation for 'onlineSavingsMsg'
Learn More

Ro 46-2005, MedChemExpress

Product Code. 16453747
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453747

Brand: Medchem Express HY19529

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ro 46-2005 is a novel synthetic non-peptide endothelin receptor antagonist, inhibits the specific binding of 125I-ET-1 to human vascular smooth muscle cells (ETA receptor) with IC50 of 220 nM.

Ro 46-2005 is a novel synthetic non-peptide endothelin receptor antagonist, inhibits the specific binding of 125I-ET-1 to human vascular smooth muscle cells (ETA receptor) with IC50 of 220 nM.

Specifications

Chemical Name or Material Ro 46-2005
CAS 150725-87-4
Color White
Purity Grade Notes Research
Molecular Formula C23H27N3O6S
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (211.18 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(C(C)(C)C)C=C1)(NC2=NC=NC(OCCO)=C2OC3=CC=CC(OC)=C3)=O
Molecular Weight (g/mol) 473.54
Formula Weight 473.54
Percent Purity 97.84%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.