Learn More
Rivaroxaban Pseudodimer, Mikromol™
Description
Rivaroxaban Pseudodimer
Specifications
Specifications
| Chemical Name or Material | Rivaroxaban Pseudodimer |
| CAS | 1632463-24-1 |
| Analyte or Component Names | Rivaroxaban Pseudodimer |
| Shipping Condition | Room Temperature |
| Molecular Formula | C38 H36 Cl2 N6 O10 S2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-Thiophenecarboxamide, 5-chloro-N-[4-[(5S)-5-[[[(5-chloro-2-thienyl)carbonyl]amino]methyl]-2-oxo-3-oxazolidinyl]phenyl]-N-[2-[2-oxo-2-[[[(5S)-2-oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]amino]ethoxy]ethyl]- |
| InChI Formula | InChI=1S/C38H36Cl2N6O10S2/c39-31-11-9-29(57-31)35(49)42-18-28-20-46(38(52)56-28)26-7-3-24(4-8-26)44(36(50)30-10-12-32(40)58-30)14-16-53-21-33(47)41-17-27-19-45(37(51)55-27)25-5-1-23(2-6-25)43-13-15-54-22-34(43)48/h1-12,27-28H,13-22H2,(H,41,47)(H,42,49)/t27-,28-/m0/s1 |
| SMILES | Clc1ccc(s1)C(=O)NC[C@H]2CN(C(=O)O2)c3ccc(cc3)N(CCOCC(=O)NC[C@H]4CN(C(=O)O4)c5ccc(cc5)N6CCOCC6=O)C(=O)c7ccc(Cl)s7 |
| IUPAC Name | 5-chloro-N-[[(5S)-3-[4-[(5-chlorothiophene-2-carbonyl)-[2-[2-oxo-2-[[(5S)-2-oxo-3-[4-(3-oxomorpholin-4-yl)phenyl]-1,3-oxazolidin-5-yl]methylamino]ethoxy]ethyl]amino]phenyl]-2-oxo-1,3-oxazolidin-5-yl]methyl]thiophene-2-carboxamide |
| Show More |