missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosomal Protein S6/RPS6 Antibody (0018) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
160.00 € - 428.00 €
Specifications
| Antigen | Ribosomal Protein S6/RPS6 |
|---|---|
| Clone | 0018 |
| Dilution | Western Blot 1:1000-1:5000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663614
|
Novus Biologicals
NBP3-23531-20ul |
20 μL |
160.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681455
|
Novus Biologicals
NBP3-23531-100ul |
100 μL |
428.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ribosomal Protein S6/RPS6 Monoclonal antibody specifically detects Ribosomal Protein S6/RPS6 in Human samples. It is validated for Western BlotSpecifications
| Ribosomal Protein S6/RPS6 | |
| Western Blot 1:1000-1:5000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, S6 | |
| The antibody is produced by immunizing rabbits with a synthetic phosphopeptide corresponding to residues around Ser240, Ser244) of Human phospho-Ribosomal Protein S6/RPS6 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0018 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, DNA Repair, mTOR Pathway, Phospho Specific, Translation Control | |
| 0.2 um filtered solution in PBS | |
| 6194 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title