missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosomal Protein S6/RPS6 Antibody (0018) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23531-100ul
This item is not returnable.
View return policy
Description
Ribosomal Protein S6/RPS6 Monoclonal antibody specifically detects Ribosomal Protein S6/RPS6 in Human samples. It is validated for Western Blot
Specifications
| Ribosomal Protein S6/RPS6 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 0018 | |
| Western Blot 1:1000-1:5000 | |
| Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, S6 | |
| The antibody is produced by immunizing rabbits with a synthetic phosphopeptide corresponding to residues around Ser240, Ser244) of Human phospho-Ribosomal Protein S6/RPS6 | |
| 100 μL | |
| Cancer, DNA Repair, mTOR Pathway, Phospho Specific, Translation Control | |
| 6194 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction