Learn More
Repaglinide Isopropyl Ester, Mikromol™
Description
Repaglinide Isopropyl Ester
Specifications
Specifications
| Chemical Name or Material | Repaglinide Isopropyl Ester |
| CAS | 2724727-17-5 |
| Analyte or Component Names | Repaglinide Isopropyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C30 H42 N2 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Repaglinide Isopropyl Ester |
| InChI Formula | InChI=1S/C30H42N2O4/c1-6-35-28-19-23(14-15-25(28)30(34)36-22(4)5)20-29(33)31-26(18-21(2)3)24-12-8-9-13-27(24)32-16-10-7-11-17-32/h8-9,12-15,19,21-22,26H,6-7,10-11,16-18,20H2,1-5H3,(H,31,33)/t26-/m0/s1 |
| SMILES | CCOc1cc(CC(=O)N[C@@H](CC(C)C)c2ccccc2N3CCCCC3)ccc1C(=O)OC(C)C |
| IUPAC Name | propan-2-yl 2-ethoxy-4-[2-[[(1S)-3-methyl-1-(2-piperidin-1-ylphenyl)butyl]amino]-2-oxoethyl]benzoate |
| Show More |