Learn More
Regadenoson, Mikromol
Product Code. 30463140
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463140 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30463140 Supplier Mikromol Supplier No. MM3772.000250
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Regadenoson, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Regadenoson
CAS 313348-27-5
Analyte or Component Names Regadenoson
Shipping Condition Room Temperature
Molecular Formula C15 H18 N8 O5
Certifications/Compliance ISO 17034
Synonym 2-[4-[(Methylamino)carbonyl]-1H-pyrazol-1-yl]adenosine, CVT 3146, Lexiscan, Regadenoson
InChI Formula InChI=1S/C15H18N8O5/c1-17-13(27)6-2-19-23(3-6)15-20-11(16)8-12(21-15)22(5-18-8)14-10(26)9(25)7(4-24)28-14/h2-3,5,7,9-10,14,24-26H,4H2,1H3,(H,17,27)(H2,16,20,21)/t7-,9-,10-,14-/m1/s1
Solution Type Neat
SMILES CNC(=O)c1cnn(c1)c2nc(N)c3ncn([C@@H]4O[C@H](CO)[C@@H](O)[C@H]4O)c3n2
IUPAC Name 1-[6-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-2-yl]-N-methylpyrazole-4-carboxamide
Molecular Weight (g/mol) 390.35
Quantity 250 mg
Formula Weight 390.14 g/mol