missing translation for 'onlineSavingsMsg'
Learn More

Refractive Index Certified Reference Material at 20 °C, 25 °C and 30 °C (Nominal RI Value 1.4023 at 25 °C), UKAS ISO 17025 / ISO 17034 Certified, in Tamper-Evident Glass Vials, 5 x 10 mL, Paragon Scientific™

Product Code. 16040290
Change view
Click to view available options
Quantity:
5 x 10 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16040290 5 x 10 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16040290 Supplier Paragon Scientific Supplier No. PSRI03K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Paragon Scientific’s range of Refractive Index Certified Reference Materials (CRMs) provide certified values for refractive index measurements at 20 °C, 25 °C and 30 °C and cover a wide range of calibration needs.

  • Refractive Index Certified Reference Materials at 20 °C, 25 °C and 30 °C
  • Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
  • Highest level of accreditation guarantee, providing the most credible certified data available worldwide
  • All measurements are fully traceable to NIST and international protocols
  • Low levels of uncertainty, ensuring maximum accuracy of data at hand and dependable results
  • Available in single 10 mL volume tamper-evident glass bottles or as a multi-pack of 5 per material (includes set of disposable pipettes)
  • 12 month shelf life
  • Manufactured in the United Kingdom

Specifications

Quantity 5 x 10 mL

Nominal values quoted. Certified Values will vary from batch to batch.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.