missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Refractive Index Certified Reference Material at 20 °C, 25 °C and 30 °C (Nominal RI Value 1.3325 at 25 °C), UKAS ISO 17025 / ISO 17034 Certified, in Tamper-Evident Glass Vials, 10 mL, Paragon Scientific™
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Description
- Refractive Index Certified Reference Materials at 20 °C, 25 °C and 30 °C
- Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
- Highest level of accreditation guarantee, providing the most credible certified data available worldwide
- All measurements are fully traceable to NIST and international protocols
- Low levels of uncertainty, ensuring maximum accuracy of data at hand and dependable results
- Available in single 10 mL volume tamper-evident glass bottles or as a multi-pack of 5 per material (includes set of disposable pipettes)
- 12 month shelf life
- Manufactured in the United Kingdom
Specifications
Specifications
| Quantity | 10 mL |
Nominal values quoted. Certified Values will vary from batch to batch.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction