missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat TLR4 / CD284 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) TLR4 / CD284, was expressed in the hosts and tags indicated.
469.00 € - 1705.00 €
Specifications
Accession Number | NP_062051.1 |
---|---|
Gene ID (Entrez) | 29260 |
Name | TLR4 / CD284 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16099141
|
enQuireBio™
QP9988-IV-100UG |
100 μg |
1705.00 €
100µg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16019151
|
enQuireBio™
QP9988-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009151
|
enQuireBio™
QP9988-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_062051.1 | |
TLR4 / CD284 Protein | |
Not determined. | |
Extracellular Domain | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
29260 | |
Research Use Only | |
Tlr4 | |
Recombinant Protein | |
E. coli | |
NPCIEVLPNI TYQCMDQNLS KIPHDIPYST KNLDLSFNPL KILRSYSFTN FSQLQWLDLS RCEIETIEDK AWHGLNQLST LVLTGNPIKS FSPGSFSGLT NLENLVAVET KMTSLEGFHI GQLISLKKLN VAHNLIHSFK LPEYFSNLTN LEHVDLSYNY IQTISVKDLQ FLRENPQVNL SLDLSLNPID SIQAQAFQGI RLHELTLRSN FNSSNVLKMC LQNMTGLHVH RLILGEFKNE RNLESFDRSV MEGLCNVSID EFRLTYINHF SDDIYNLNCL ANISAMSFTG VHIKHIADVP RHFKWQSLSI IRCHLKPFPK LSLPFLKSWT LTTNREDISF GQLALPSLRY LDLSRNAMSF RGCCSYSDFG TNNLKYLDLS FNGVILMSAN FMGLEELEYL DFQHSTLKKV TEFSVFLSLE KLLYLDISYT NTKIDFDGIF LGLISLNTLK MAGNSFKDNT LSNVFTNTTN LTFLDLSKCQ LEQISRGVFD TLYRLQLLNM SHNNLLFLDP SHYKQLYSLR TLDCSFNRIE TSKGILQHFP KSLAVFNLTN NSVACICEYQ NFLQWVKDQK MFLVNVEQMK CASPIDMKAS LVLDFTNSTC YIYKTIISVS VVS | |
Greater than 85% as determined by SDS-PAGE. |