missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat TLR4 / CD284 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) TLR4 / CD284, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9988-iv-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_062051.1 | |
TLR4 / CD284 Protein | |
Research Use Only | |
Tlr4 | |
Recombinant Protein | |
E. coli | |
NPCIEVLPNI TYQCMDQNLS KIPHDIPYST KNLDLSFNPL KILRSYSFTN FSQLQWLDLS RCEIETIEDK AWHGLNQLST LVLTGNPIKS FSPGSFSGLT NLENLVAVET KMTSLEGFHI GQLISLKKLN VAHNLIHSFK LPEYFSNLTN LEHVDLSYNY IQTISVKDLQ FLRENPQVNL SLDLSLNPID SIQAQAFQGI RLHELTLRSN FNSSNVLKMC LQNMTGLHVH RLILGEFKNE RNLESFDRSV MEGLCNVSID EFRLTYINHF SDDIYNLNCL ANISAMSFTG VHIKHIADVP RHFKWQSLSI IRCHLKPFPK LSLPFLKSWT LTTNREDISF GQLALPSLRY LDLSRNAMSF RGCCSYSDFG TNNLKYLDLS FNGVILMSAN FMGLEELEYL DFQHSTLKKV TEFSVFLSLE KLLYLDISYT NTKIDFDGIF LGLISLNTLK MAGNSFKDNT LSNVFTNTTN LTFLDLSKCQ LEQISRGVFD TLYRLQLLNM SHNNLLFLDP SHYKQLYSLR TLDCSFNRIE TSKGILQHFP KSLAVFNLTN NSVACICEYQ NFLQWVKDQK MFLVNVEQMK CASPIDMKAS LVLDFTNSTC YIYKTIISVS VVS | |
Greater than 85% as determined by SDS-PAGE. |
29260 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |