missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Oncomodulin Protein
A DNA sequence encoding the Mus musculus (Mouse) Oncomodulin, was expressed in the hosts and tags indicated.
287.00 € - 3175.00 €
Specifications
Accession Number | NP_149028.2 |
---|---|
Gene ID (Entrez) | 18261 |
Name | Oncomodulin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003360
|
enQuireBio™
QP6444-YE-100UG |
100 μg |
920.00 €
100µg |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043360
|
enQuireBio™
QP6444-YE-500UG |
500 μg |
2650.00 €
500µg |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013360
|
enQuireBio™
QP6444-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033360
|
enQuireBio™
QP6444-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053360
|
enQuireBio™
QP6444-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023360
|
enQuireBio™
QP6444-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_149028.2 | |
Oncomodulin Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
18261 | |
Research Use Only | |
Ocm | |
Recombinant Protein | |
Yeast | |
SITDILSADD IAAALQECQD PDTFEPQKFF QTSGLSKMSA SQLKDIFQFI DNDQSGYLDE DELKYFLQRF QSDARELTES ETKSLMDAAD NDGDGKIGAD EFQEMVHS | |
Greater than 90% as determined by SDS-PAGE. |