missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Oncomodulin Protein
A DNA sequence encoding the Mus musculus (Mouse) Oncomodulin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6444-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_149028.2 | |
Oncomodulin Protein | |
Research Use Only | |
Ocm | |
Recombinant Protein | |
Yeast | |
SITDILSADD IAAALQECQD PDTFEPQKFF QTSGLSKMSA SQLKDIFQFI DNDQSGYLDE DELKYFLQRF QSDARELTES ETKSLMDAAD NDGDGKIGAD EFQEMVHS | |
Greater than 90% as determined by SDS-PAGE. |
18261 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |