missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Mucosal addressin cell adhesion molecule 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Mucosal addressin cell adhesion molecule 1, was expressed in the hosts and tags indicated.
245.00 € - 3405.00 €
Specifications
Accession Number | XP_006513356.1 |
---|---|
Gene ID (Entrez) | 17123 |
Name | Mucosal addressin cell adhesion molecule 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16080191
|
enQuireBio™
QP7689-EC-100UG |
100 μg |
770.00 €
100µg |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16000201
|
enQuireBio™
QP7689-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090191
|
enQuireBio™
QP7689-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16010201
|
enQuireBio™
QP7689-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020201
|
enQuireBio™
QP7689-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030201
|
enQuireBio™
QP7689-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
XP_006513356.1 | |
Mucosal addressin cell adhesion molecule 1 Protein | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
17123 | |
Research Use Only | |
Madcam1 | |
Recombinant Protein | |
E. coli | |
QSFQVNPPES EVAVAMGTSL QITCSMSCDE GVARVHWRGL DTSLGSVQTL PGSSILSVRG MLSDTGTPVC VGSCGSRSFQ HSVKILVYAF PDQLVVSPEF LVPGQDQVVS CTAHNIWPAD PNSLSFALLL GEQRLEGAQA LEPEQEEEIQ EAEGTPLFRM TQRWRLPSLG TPAPPALHCQ VTMQLPKLVL THRKEIPVLQ SQTSPKPPNT TSAEPYILTS SSTAEAVSTG LNITTLPSAP PYPKLSPRTL SSEGPCRPKI HQDLEAGWEL LCEASCGPGV TVRWTLAPGD LATYHKREAG AQAWLSVLPP GPMVEGWFQC RQDPGGEVTN LYVPGQVTPN SSS | |
Greater than 90% as determined by SDS-PAGE. |