missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Mucosal addressin cell adhesion molecule 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Mucosal addressin cell adhesion molecule 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7689-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
XP_006513356.1 | |
Mucosal addressin cell adhesion molecule 1 Protein | |
Research Use Only | |
Madcam1 | |
Recombinant Protein | |
E. coli | |
QSFQVNPPES EVAVAMGTSL QITCSMSCDE GVARVHWRGL DTSLGSVQTL PGSSILSVRG MLSDTGTPVC VGSCGSRSFQ HSVKILVYAF PDQLVVSPEF LVPGQDQVVS CTAHNIWPAD PNSLSFALLL GEQRLEGAQA LEPEQEEEIQ EAEGTPLFRM TQRWRLPSLG TPAPPALHCQ VTMQLPKLVL THRKEIPVLQ SQTSPKPPNT TSAEPYILTS SSTAEAVSTG LNITTLPSAP PYPKLSPRTL SSEGPCRPKI HQDLEAGWEL LCEASCGPGV TVRWTLAPGD LATYHKREAG AQAWLSVLPP GPMVEGWFQC RQDPGGEVTN LYVPGQVTPN SSS | |
Greater than 90% as determined by SDS-PAGE. |
17123 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |