missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Kallikrein-14 Protein
A DNA sequence encoding the Mus musculus (Mouse) Kallikrein-14, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9831-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_777355.1 | |
Kallikrein-14 Protein | |
Research Use Only | |
Klk14 | |
Recombinant Protein | |
Yeast | |
IIGGYRCVRN SQPWQVALQA GPGHRFLCGG VLLSDQWVIT AAHCARPILH VALGKHNIRR WEATQQVVRV ARQVPHPQYQ PQAHDNDLML LKLQKKVRLG RAVKTISVAS SCASPGTPCR VSGWGTIASP IARYPTALQC VNVNIMSEQA CHRAYPGIIT SGMVCAGVPE GGKDSCQGDS GGPLVCGGQL QGLVSWGMER CAMPGYPGVY ANLCNYHSWI QRTMQSN | |
Greater than 84% as determined by SDS-PAGE. |
317653 | |
1 mg | |
Not determined. | |
Full. Length. of. Mature. Protein | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |