missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Kallikrein-14 Protein
A DNA sequence encoding the Mus musculus (Mouse) Kallikrein-14, was expressed in the hosts and tags indicated.
245.00 € - 3405.00 €
Specifications
Accession Number | NP_777355.1 |
---|---|
Gene ID (Entrez) | 317653 |
Name | Kallikrein-14 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16068811
|
enQuireBio™
QP9831-YE-100UG |
100 μg |
770.00 €
100µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088811
|
enQuireBio™
QP9831-YE-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078811
|
enQuireBio™
QP9831-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098811
|
enQuireBio™
QP9831-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008821
|
enQuireBio™
QP9831-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018821
|
enQuireBio™
QP9831-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_777355.1 | |
Kallikrein-14 Protein | |
Not determined. | |
Full. Length. of. Mature. Protein | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
317653 | |
Research Use Only | |
Klk14 | |
Recombinant Protein | |
Yeast | |
IIGGYRCVRN SQPWQVALQA GPGHRFLCGG VLLSDQWVIT AAHCARPILH VALGKHNIRR WEATQQVVRV ARQVPHPQYQ PQAHDNDLML LKLQKKVRLG RAVKTISVAS SCASPGTPCR VSGWGTIASP IARYPTALQC VNVNIMSEQA CHRAYPGIIT SGMVCAGVPE GGKDSCQGDS GGPLVCGGQL QGLVSWGMER CAMPGYPGVY ANLCNYHSWI QRTMQSN | |
Greater than 84% as determined by SDS-PAGE. |