missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD90 / THY-1 Protein

A DNA sequence encoding the Mus musculus (Mouse) CD90 / THY-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6789-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055870

  • 3405.00 € / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_033408.1
CD90 / THY-1 Protein
Research Use Only
Thy1
Recombinant Protein
E. coli
QKVTSLTACL VNQNLRLDCR HENNTKDNSI QHEFSLTREK RKHVLSGTLG IPEHTYRSRV TLSNQPYIKV LTLANFTTKD EGDYFCELQV SGANPMSSNK SISVYRDKLV KC
Greater than 90% as determined by SDS-PAGE.
21838
1 mg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions