missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD90 / THY-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) CD90 / THY-1, was expressed in the hosts and tags indicated.
309.00 € - 3405.00 €
Specifications
Accession Number | NP_033408.1 |
---|---|
Gene ID (Entrez) | 21838 |
Name | CD90 / THY-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035870
|
enQuireBio™
QP6789-EC-100UG |
100 μg |
985.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16055870
|
enQuireBio™
QP6789-EC-1MG |
1 mg |
3405.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045870
|
enQuireBio™
QP6789-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065870
|
enQuireBio™
QP6789-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075870
|
enQuireBio™
QP6789-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085870
|
enQuireBio™
QP6789-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_033408.1 | |
CD90 / THY-1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
21838 | |
Research Use Only | |
Thy1 | |
Recombinant Protein | |
E. coli | |
QKVTSLTACL VNQNLRLDCR HENNTKDNSI QHEFSLTREK RKHVLSGTLG IPEHTYRSRV TLSNQPYIKV LTLANFTTKD EGDYFCELQV SGANPMSSNK SISVYRDKLV KC | |
Greater than 90% as determined by SDS-PAGE. |