missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Arid5a Protein
A DNA sequence encoding the Mus musculus (Mouse) Arid5a, was expressed in the hosts and tags indicated.
287.00 € - 3175.00 €
Specifications
Accession Number | NP_001165676.1 |
---|---|
Gene ID (Entrez) | 214855 |
Name | Arid5a Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028731
|
enQuireBio™
QP9789-YE-100UG |
100 μg |
920.00 €
100µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16068731
|
enQuireBio™
QP9789-YE-500UG |
500 μg |
2650.00 €
500µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038731
|
enQuireBio™
QP9789-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058731
|
enQuireBio™
QP9789-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078731
|
enQuireBio™
QP9789-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048731
|
enQuireBio™
QP9789-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001165676.1 | |
Arid5a Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
214855 | |
Research Use Only | |
Arid5a | |
Recombinant Protein | |
Yeast | |
MAAPPAKGNT EQSEEGDLPQ LPVSPKPDDE QSRSQSPTQL QDSPEAGGEQ EEEQAFLVSL YKFMKERHTP IERVPHLGFK QINLWKIYKA VEKLGAYELV TGRRLWKNVY DELGGSPGST SAATCTRRHY ERLVLPYVRH LKGEDDKPLP PTKPRKQYKM AKELRGDDGT TEKLKKAKDS EERRVEQTTP GKTKSDATGQ TQLPCQGSSR DSTEQLGPVS GPSPPLTGAS SCPEAYKRLL SSFYCKGAHG IMSPLAKKKL LAQVSKAEAL QCQEEGCRHG ARSPNKDIQD SPQNLRGPAE NSEHQLTPRE GLQAPGGSTR MEAQVGPCPT APMFSGCFHA YPTEVLKPVS QHPRDFFSGL KDRVLLGPPG KEEGPTTKES HLVWGGDANH PSAFHKGSTR KRSFYPKPKA CWVSPMAKVP TERPGAPSPH PSSPGLGSKR GLEEEGFAHG GKKLRAVSPF LKEVDSKETG GKPAAPGLAV SCLLGPTPGP TPPEAYRGTM LRCPLNFTGS ADPLKGQASL PFSPLVIPAF PAHLLATTGS SPMAASLMHF PPTPYDAVLR NRLGPASSAW HMPPVTTYAA PHFFHLNTKL | |
Greater than 90% as determined by SDS-PAGE. |