missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Arid5a Protein
A DNA sequence encoding the Mus musculus (Mouse) Arid5a, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9789-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001165676.1 | |
Arid5a Protein | |
Research Use Only | |
Arid5a | |
Recombinant Protein | |
Yeast | |
MAAPPAKGNT EQSEEGDLPQ LPVSPKPDDE QSRSQSPTQL QDSPEAGGEQ EEEQAFLVSL YKFMKERHTP IERVPHLGFK QINLWKIYKA VEKLGAYELV TGRRLWKNVY DELGGSPGST SAATCTRRHY ERLVLPYVRH LKGEDDKPLP PTKPRKQYKM AKELRGDDGT TEKLKKAKDS EERRVEQTTP GKTKSDATGQ TQLPCQGSSR DSTEQLGPVS GPSPPLTGAS SCPEAYKRLL SSFYCKGAHG IMSPLAKKKL LAQVSKAEAL QCQEEGCRHG ARSPNKDIQD SPQNLRGPAE NSEHQLTPRE GLQAPGGSTR MEAQVGPCPT APMFSGCFHA YPTEVLKPVS QHPRDFFSGL KDRVLLGPPG KEEGPTTKES HLVWGGDANH PSAFHKGSTR KRSFYPKPKA CWVSPMAKVP TERPGAPSPH PSSPGLGSKR GLEEEGFAHG GKKLRAVSPF LKEVDSKETG GKPAAPGLAV SCLLGPTPGP TPPEAYRGTM LRCPLNFTGS ADPLKGQASL PFSPLVIPAF PAHLLATTGS SPMAASLMHF PPTPYDAVLR NRLGPASSAW HMPPVTTYAA PHFFHLNTKL | |
Greater than 90% as determined by SDS-PAGE. |
214855 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |