missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Uncharacterized protein C19orf48 Protein
A DNA sequence encoding the Homo sapiens (Human) Uncharacterized protein C19orf48, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7738-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001277078.1 | |
Uncharacterized protein C19orf48 Protein | |
Research Use Only | |
C19orf48 | |
Recombinant Protein | |
E. coli | |
MTVLEAVLEI QAITGSRLLS MVPGPARPPG SCWDPTQCTR TWLLSHTPRR RWISGLPRAS CRLGEEPPPL PYCDQAYGEE LSIRHRETWA WLSRTDTAWP GAPGVKQARI LGELLLV | |
Greater than 80% as determined by SDS-PAGE. |
84798 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |