Learn More
enQuireBio™ Recombinant Human Uncharacterized protein C19orf48 Protein
A DNA sequence encoding the Homo sapiens (Human) Uncharacterized protein C19orf48, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001277078.1 |
---|---|
Gene ID (Entrez) | 84798 |
Name | Uncharacterized protein C19orf48 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16080511
|
enQuireBio™
QP7738-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16000521
|
enQuireBio™
QP7738-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090511
|
enQuireBio™
QP7738-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16010521
|
enQuireBio™
QP7738-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020521
|
enQuireBio™
QP7738-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030521
|
enQuireBio™
QP7738-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001277078.1 | |
Uncharacterized protein C19orf48 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
84798 | |
Research Use Only | |
C19orf48 | |
Recombinant Protein | |
E. coli | |
MTVLEAVLEI QAITGSRLLS MVPGPARPPG SCWDPTQCTR TWLLSHTPRR RWISGLPRAS CRLGEEPPPL PYCDQAYGEE LSIRHRETWA WLSRTDTAWP GAPGVKQARI LGELLLV | |
Greater than 80% as determined by SDS-PAGE. |