missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human THAP domain-containing protein 11 Protein
A DNA sequence encoding the Homo sapiens (Human) THAP domain-containing protein 11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7939-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_065190.2 | |
THAP domain-containing protein 11 Protein | |
Research Use Only | |
THAP11 | |
Recombinant Protein | |
E. coli | |
MPGFTCCVPG CYNNSHRDKA LHFYTFPKDA ELRRLWLKNV SRAGVSGCFS TFQPTTGHRL CSVHFQGGRK TYTVRVPTIF PLRGVNERKV ARRPAGAAAA RRRQQQQQQQ QQQQQQQQQQ QQQQQQQQQQ QQSSPSASTA QTAQLQPNLV SASAAVLLTL QATVDSSQAP GSVQPAPITP TGEDVKPIDL TVQVEFAAAE GAAAAAAASE LQAATAGLEA AECPMGPQLV VVGEEGFPDT GSDHSYSLSS GTTEEELLRK LNEQRDILAL MEVKMKEMKG SIRHLRLTEA KLREELREKD RLLAMAVIRK KHG | |
Greater than 90% as determined by SDS-PAGE. |
57215 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |