missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human THAP domain-containing protein 11 Protein
A DNA sequence encoding the Homo sapiens (Human) THAP domain-containing protein 11, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_065190.2 |
---|---|
Gene ID (Entrez) | 57215 |
Name | THAP domain-containing protein 11 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16071721
|
enQuireBio™
QP7939-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16091721
|
enQuireBio™
QP7939-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16081721
|
enQuireBio™
QP7939-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001731
|
enQuireBio™
QP7939-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011731
|
enQuireBio™
QP7939-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021731
|
enQuireBio™
QP7939-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_065190.2 | |
THAP domain-containing protein 11 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
57215 | |
Research Use Only | |
THAP11 | |
Recombinant Protein | |
E. coli | |
MPGFTCCVPG CYNNSHRDKA LHFYTFPKDA ELRRLWLKNV SRAGVSGCFS TFQPTTGHRL CSVHFQGGRK TYTVRVPTIF PLRGVNERKV ARRPAGAAAA RRRQQQQQQQ QQQQQQQQQQ QQQQQQQQQQ QQSSPSASTA QTAQLQPNLV SASAAVLLTL QATVDSSQAP GSVQPAPITP TGEDVKPIDL TVQVEFAAAE GAAAAAAASE LQAATAGLEA AECPMGPQLV VVGEEGFPDT GSDHSYSLSS GTTEEELLRK LNEQRDILAL MEVKMKEMKG SIRHLRLTEA KLREELREKD RLLAMAVIRK KHG | |
Greater than 90% as determined by SDS-PAGE. |