missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human T-box transcription factor TBX15 Protein
A DNA sequence encoding the Homo sapiens (Human) T-box transcription factor TBX15, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001317606.1 |
---|---|
Gene ID (Entrez) | 6913 |
Name | T-box transcription factor TBX15 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16071481
|
enQuireBio™
QP7900-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16091481
|
enQuireBio™
QP7900-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16081481
|
enQuireBio™
QP7900-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001491
|
enQuireBio™
QP7900-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011491
|
enQuireBio™
QP7900-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021491
|
enQuireBio™
QP7900-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001317606.1 | |
T-box transcription factor TBX15 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6913 | |
Research Use Only | |
TBX15 | |
Recombinant Protein | |
E. coli | |
MSSMEEIQVE LQCADLWKRF HDIGTEMIIT KAGRRMFPAM RVKITGLDPH QQYYIAMDIV PVDNKRYRYV YHSSKWMVAG NADSPVPPRV YIHPDSLASG DTWMRQVVSF DKLKLTNNEL DDQGHIILHS MHKYQPRVHV IRKDFSSDLS PTKPVPVGDG VKTFNFPETV FTTVTAYQNQ QITRLKIDRN PFAKGFRDSG RNRTGLEAIM ETYAFWRPPV RTLTFEDFTT MQKQQGGSTG TSPTTSSTGT PSPSASSHLL SPSCSPPTFH LAPNTFNVGC RESQLCNLNL SDYPPCARSN MAALQSYPGL SDSGYNRLQS GTTSATQPSE TFMPQRTPSL ISGIPTPPSL PGNSKMEAYG GQLGSFPTSQ FQYVMQAGNA ASSSSSPHMF GGSHMQQSSY NAFSLHNPYN LYGYNFPTSP RLAASPEKLS ASQSTLLCSS PSNGAFGERQ YLPSGMEHSM HMISPSPNNQ QATNTCDGRQ YGAVPGSSSQ MSVH | |
Greater than 90% as determined by SDS-PAGE. |