missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human T-box transcription factor TBX15 Protein
A DNA sequence encoding the Homo sapiens (Human) T-box transcription factor TBX15, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7900-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001317606.1 | |
T-box transcription factor TBX15 Protein | |
Research Use Only | |
TBX15 | |
Recombinant Protein | |
E. coli | |
MSSMEEIQVE LQCADLWKRF HDIGTEMIIT KAGRRMFPAM RVKITGLDPH QQYYIAMDIV PVDNKRYRYV YHSSKWMVAG NADSPVPPRV YIHPDSLASG DTWMRQVVSF DKLKLTNNEL DDQGHIILHS MHKYQPRVHV IRKDFSSDLS PTKPVPVGDG VKTFNFPETV FTTVTAYQNQ QITRLKIDRN PFAKGFRDSG RNRTGLEAIM ETYAFWRPPV RTLTFEDFTT MQKQQGGSTG TSPTTSSTGT PSPSASSHLL SPSCSPPTFH LAPNTFNVGC RESQLCNLNL SDYPPCARSN MAALQSYPGL SDSGYNRLQS GTTSATQPSE TFMPQRTPSL ISGIPTPPSL PGNSKMEAYG GQLGSFPTSQ FQYVMQAGNA ASSSSSPHMF GGSHMQQSSY NAFSLHNPYN LYGYNFPTSP RLAASPEKLS ASQSTLLCSS PSNGAFGERQ YLPSGMEHSM HMISPSPNNQ QATNTCDGRQ YGAVPGSSSQ MSVH | |
Greater than 90% as determined by SDS-PAGE. |
6913 | |
1 mg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |