missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ST3GAL3 Protein
A DNA sequence encoding the Homo sapiens (Human) ST3GAL3, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001257388.1 |
---|---|
Gene ID (Entrez) | 6487 |
Name | ST3GAL3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059140
|
enQuireBio™
QP7519-EC-100UG |
100 μg |
525.00 €
100µg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16079140
|
enQuireBio™
QP7519-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16069140
|
enQuireBio™
QP7519-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089140
|
enQuireBio™
QP7519-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099140
|
enQuireBio™
QP7519-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009150
|
enQuireBio™
QP7519-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001257388.1 | |
ST3GAL3 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6487 | |
Research Use Only | |
ST3GAL3 | |
Recombinant Protein | |
E. coli | |
KLHLLQWEED SNSVVLSFDS AGQTLGSEYD RLGFLLNLDS KLPAELATKY ANFSEGACKP GYASALMTAI FPRFSKPAPM FLDDSFRKWA RIREFVPPFG IKGQDNLIKA ILSVTKEYRL TPALDSLRCR RCIIVGNGGV LANKSLGSRI DDYDIVVRLN SAPVKGFEKD VGSKTTLRIT YPEGAMQRPE QYERDSLFVL AGFKWQDFKW LKYIVYKERV SASDGFWKSV ATRVPKEPPE IRILNPYFIQ EAAFTLIGLP FNNGLMGRGN IPTLGSVAVT MALHGCDEVA VAGFGYDMST PNAPLHYYET VRMAAIKESW THNIQREKEF LRKLVKARVI TDLSSGI | |
Greater than 90% as determined by SDS-PAGE. |