missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ST3GAL3 Protein
A DNA sequence encoding the Homo sapiens (Human) ST3GAL3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7519-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001257388.1 | |
ST3GAL3 Protein | |
Research Use Only | |
ST3GAL3 | |
Recombinant Protein | |
E. coli | |
KLHLLQWEED SNSVVLSFDS AGQTLGSEYD RLGFLLNLDS KLPAELATKY ANFSEGACKP GYASALMTAI FPRFSKPAPM FLDDSFRKWA RIREFVPPFG IKGQDNLIKA ILSVTKEYRL TPALDSLRCR RCIIVGNGGV LANKSLGSRI DDYDIVVRLN SAPVKGFEKD VGSKTTLRIT YPEGAMQRPE QYERDSLFVL AGFKWQDFKW LKYIVYKERV SASDGFWKSV ATRVPKEPPE IRILNPYFIQ EAAFTLIGLP FNNGLMGRGN IPTLGSVAVT MALHGCDEVA VAGFGYDMST PNAPLHYYET VRMAAIKESW THNIQREKEF LRKLVKARVI TDLSSGI | |
Greater than 90% as determined by SDS-PAGE. |
6487 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |