Learn More
enQuireBio™ Recombinant Human Ribonuclease P protein subunit p20 Protein
A DNA sequence encoding the Homo sapiens (Human) Ribonuclease P protein subunit p20, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_005828.2 |
---|---|
Gene ID (Entrez) | 10248 |
Name | Ribonuclease P protein subunit p20 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083840
|
enQuireBio™
QP6515-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003850
|
enQuireBio™
QP6515-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093840
|
enQuireBio™
QP6515-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013850
|
enQuireBio™
QP6515-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023850
|
enQuireBio™
QP6515-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033850
|
enQuireBio™
QP6515-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005828.2 | |
Ribonuclease P protein subunit p20 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10248 | |
Research Use Only | |
POP7 | |
Recombinant Protein | |
E. coli | |
MAENREPRGA VEAELDPVEY TLRKRLPSRL PRRPNDIYVN MKTDFKAQLA RCQKLLDGGA RGQNACSEIY IHGLGLAINR AINIALQLQA GSFGSLQVAA NTSTVELVDE LEPETDTREP LTRIRNNSAI HIRVFRVTPK | |
Greater than 80% as determined by SDS-PAGE. |