Learn More
enQuireBio™ Recombinant Human Ribonuclease P protein subunit p20 Protein
A DNA sequence encoding the Homo sapiens (Human) Ribonuclease P protein subunit p20, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6515-ec-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16003850
  • 2385.00 € / 1mg
Estimated Shipment: 08-07-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Specifications
NP_005828.2
Ribonuclease P protein subunit p20 Protein
Research Use Only
POP7
Recombinant Protein
E. coli
MAENREPRGA VEAELDPVEY TLRKRLPSRL PRRPNDIYVN MKTDFKAQLA RCQKLLDGGA RGQNACSEIY IHGLGLAINR AINIALQLQA GSFGSLQVAA NTSTVELVDE LEPETDTREP LTRIRNNSAI HIRVFRVTPK
Greater than 80% as determined by SDS-PAGE.
10248
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol