missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ragulator complex protein LAMTOR5 Protein
A DNA sequence encoding the Homo sapiens (Human) Ragulator complex protein LAMTOR5, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_006393.2 |
---|---|
Gene ID (Entrez) | 10542 |
Name | Ragulator complex protein LAMTOR5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091160
|
enQuireBio™
QP6147-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16021170
|
enQuireBio™
QP6147-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071160
|
enQuireBio™
QP6147-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001170
|
enQuireBio™
QP6147-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011170
|
enQuireBio™
QP6147-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081160
|
enQuireBio™
QP6147-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006393.2 | |
Ragulator complex protein LAMTOR5 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10542 | |
Research Use Only | |
HBXIP | |
Recombinant Protein | |
E. coli | |
MEATLEQHLE DTMKNPSIVG VLCTDSQGLN LGCRGTLSDE HAGVISVLAQ QAAKLTSDPT DIPVVCLESD NGNIMIQKHD GITVAVHKMA S | |
Greater than 80% as determined by SDS-PAGE. |