missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ragulator complex protein LAMTOR5 Protein
A DNA sequence encoding the Homo sapiens (Human) Ragulator complex protein LAMTOR5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6147-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006393.2 | |
Ragulator complex protein LAMTOR5 Protein | |
Research Use Only | |
HBXIP | |
Recombinant Protein | |
E. coli | |
MEATLEQHLE DTMKNPSIVG VLCTDSQGLN LGCRGTLSDE HAGVISVLAQ QAAKLTSDPT DIPVVCLESD NGNIMIQKHD GITVAVHKMA S | |
Greater than 80% as determined by SDS-PAGE. |
10542 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |