missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein MIS12 homolog Protein

A DNA sequence encoding the Homo sapiens (Human) Protein MIS12 homolog, was expressed in the hosts and tags indicated.

160.00 € - 2385.00 €

Specifications

Accession Number NP_001245146.1
Gene ID (Entrez) 79003
Name Protein MIS12 homolog Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16072861
View Documents Promotion Details
enQuireBio™
QP8140-EC-100UG
100 μg
530.00 €
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16092861
View Documents Promotion Details
enQuireBio™
QP8140-EC-1MG
1 mg
2385.00 €
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16082861
View Documents Promotion Details
enQuireBio™
QP8140-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16002871
View Documents Promotion Details
enQuireBio™
QP8140-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16012871
View Documents Promotion Details
enQuireBio™
QP8140-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16022871
View Documents Promotion Details
enQuireBio™
QP8140-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001245146.1
Protein MIS12 homolog Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
79003
Research Use Only
MIS12
Recombinant Protein
E. coli
MSVDPMTYEA QFFGFTPQTC MLRIYIAFQD YLFEVMQAVE QVILKKLDGI PDCDISPVQI RKCTEKFLCF MKGHFDNLFS KMEQLFLQLI LRIPSNILLP EDKCKETPYS EEDFQHLQKE IEQLQEKYKT ELCTKQALLA ELEEQKIVQA KLKQTLTFFD ELHNVGRDHG TSDFRESLVS LVQNSRKLQN IRDNVEKESK RLKIS
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications