missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein MIS12 homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Protein MIS12 homolog, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8140-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001245146.1 | |
Protein MIS12 homolog Protein | |
Research Use Only | |
MIS12 | |
Recombinant Protein | |
E. coli | |
MSVDPMTYEA QFFGFTPQTC MLRIYIAFQD YLFEVMQAVE QVILKKLDGI PDCDISPVQI RKCTEKFLCF MKGHFDNLFS KMEQLFLQLI LRIPSNILLP EDKCKETPYS EEDFQHLQKE IEQLQEKYKT ELCTKQALLA ELEEQKIVQA KLKQTLTFFD ELHNVGRDHG TSDFRESLVS LVQNSRKLQN IRDNVEKESK RLKIS | |
Greater than 90% as determined by SDS-PAGE. |
79003 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |