missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein-arginine deiminase type-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein-arginine deiminase type-3, was expressed in the hosts and tags indicated.
287.00 € - 4075.00 €
Specifications
Accession Number | NP_057317.2 |
---|---|
Gene ID (Entrez) | 51702 |
Name | Protein-arginine deiminase type-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16069021
|
enQuireBio™
QP9930-YE-100UG |
100 μg |
920.00 €
100µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16089021
|
enQuireBio™
QP9930-YE-1MG |
1 mg |
4075.00 €
1mg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16079021
|
enQuireBio™
QP9930-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099021
|
enQuireBio™
QP9930-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009031
|
enQuireBio™
QP9930-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16019031
|
enQuireBio™
QP9930-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Especificaciones
NP_057317.2 | |
Protein-arginine deiminase type-3 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
51702 | |
Research Use Only | |
PADI3 | |
Recombinant Protein | |
Yeast | |
MSLQRIVRVS LEHPTSAVCV AGVETLVDIY GSVPEGTEMF EVYGTPGVDI YISPNMERGR ERADTRRWRF DATLEIIVVM NSPSNDLNDS HVQISYHSSH EPLPLAYAVL YLTCVDISLD CDLNCEGRQD RNFVDKRQWV WGPSGYGGIL LVNCDRDDPS CDVQDNCDQH VHCLQDLEDM SVMVLRTQGP AALFDDHKLV LHTSSYDAKR AQVFHICGPE DVCEAYRHVL GQDKVSYEVP RLHGDEERFF VEGLSFPDAG FTGLISFHVT LLDDSNEDFS ASPIFTDTVV FRVAPWIMTP STLPPLEVYV CRVRNNTCFV DAVAELARKA GCKLTICPQA ENRNDRWIQD EMELGYVQAP HKTLPVVFDS PRNGELQDFP YKRILGPDFG YVTREPRDRS VSGLDSFGNL EVSPPVVANG KEYPLGRILI GGNLPGSSGR RVTQVVRDFL HAQKVQPPVE LFVDWLAVGH VDEFLSFVPA PDGKGFRMLL ASPGACFKLF QEKQKCGHGR ALLFQGVVDD EQVKTISINQ VLSNKDLINY NKFVQSCIDW NREVLKRELG LAECDIIDIP QLFKTERKKA TAFFPDLVNM LVLGKHLGIP KPFGPIINGC CCLEEKVRSL LEPLGLHCTF IDDFTPYHML HGEVHCGTNV CRKPFSFKWW NMVP | |
Greater than 90% as determined by SDS-PAGE. |