missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein-arginine deiminase type-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein-arginine deiminase type-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9930-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_057317.2 | |
Protein-arginine deiminase type-3 Protein | |
Research Use Only | |
PADI3 | |
Recombinant Protein | |
Yeast | |
MSLQRIVRVS LEHPTSAVCV AGVETLVDIY GSVPEGTEMF EVYGTPGVDI YISPNMERGR ERADTRRWRF DATLEIIVVM NSPSNDLNDS HVQISYHSSH EPLPLAYAVL YLTCVDISLD CDLNCEGRQD RNFVDKRQWV WGPSGYGGIL LVNCDRDDPS CDVQDNCDQH VHCLQDLEDM SVMVLRTQGP AALFDDHKLV LHTSSYDAKR AQVFHICGPE DVCEAYRHVL GQDKVSYEVP RLHGDEERFF VEGLSFPDAG FTGLISFHVT LLDDSNEDFS ASPIFTDTVV FRVAPWIMTP STLPPLEVYV CRVRNNTCFV DAVAELARKA GCKLTICPQA ENRNDRWIQD EMELGYVQAP HKTLPVVFDS PRNGELQDFP YKRILGPDFG YVTREPRDRS VSGLDSFGNL EVSPPVVANG KEYPLGRILI GGNLPGSSGR RVTQVVRDFL HAQKVQPPVE LFVDWLAVGH VDEFLSFVPA PDGKGFRMLL ASPGACFKLF QEKQKCGHGR ALLFQGVVDD EQVKTISINQ VLSNKDLINY NKFVQSCIDW NREVLKRELG LAECDIIDIP QLFKTERKKA TAFFPDLVNM LVLGKHLGIP KPFGPIINGC CCLEEKVRSL LEPLGLHCTF IDDFTPYHML HGEVHCGTNV CRKPFSFKWW NMVP | |
Greater than 90% as determined by SDS-PAGE. |
51702 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |