missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PD1 / PDCD1 / CD279 Protein

A DNA sequence encoding the Homo sapiens (Human) PD1 / PDCD1 / CD279, was expressed in the hosts and tags indicated.

160.00 € - 2385.00 €

Specifications

Accession Number NP_005009.2
Gene ID (Entrez) 5133
Name PD1 / PDCD1 / CD279 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16069490
View Documents Promotion Details
enQuireBio™
QP7574-EC-100UG
100 μg
530.00 €
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 09-07-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16089490
View Documents Promotion Details
enQuireBio™
QP7574-EC-1MG
1 mg
2385.00 €
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 09-07-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16079490
View Documents Promotion Details
enQuireBio™
QP7574-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16099490
View Documents Promotion Details
enQuireBio™
QP7574-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16009500
View Documents Promotion Details
enQuireBio™
QP7574-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16019500
View Documents Promotion Details
enQuireBio™
QP7574-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_005009.2
PD1 / PDCD1 / CD279 Protein
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
5133
Research Use Only
PDCD1
Recombinant Protein
E. coli
PGWFLDSPDR PWNPPTFSPA LLVVTEGDNA TFTCSFSNTS ESFVLNWYRM SPSNQTDKLA AFPEDRSQPG QDCRFRVTQL PNGRDFHMSV VRARRNDSGT YLCGAISLAP KAQIKESLRA ELRVTERRAE VPTAHPSPSP RPAGQFQTLV
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications